Learn More
Hampton Research PEG/ION SCREEN NO46 185ML
SDP
Catalog No. 501869
Encompass
HR2-922-46/PEG/Ion Screen #46 (D10), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-9 Supplier Hampton Research Supplier No. HR292246
HR2-922-46/PEG/Ion Screen #46 (D10), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.