Learn More
Hampton Research PEG/ION SCREEN NO48 185ML
SDP
Supplier:  Hampton Research HR292248
Encompass
HR2-922-48/PEG/Ion Screen #48 (D12), 185 ml - 0.2 M Ammonium citrate dibasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-187-1