Learn More
System Biosciences LLC PEG-IT VIRUS PRECIPITATION PEG-it Virus Precipitation Solution = 250mL
SDP
Catalog No. NC9846201
Encompass_Preferred
Easily concentrate lentivirus particles for ultra-high titers. Used in over three-hundred citations, PEG-it™ Virus Concentration Reagent enables easy concentration of pseudoviral particles for achieving ultra-high titers. It’s great for concentrating pseudoviral particles even from large volumes of medium by removing the need for ultracentrifugation. Simply add PEG-it to the collected medium, incubate overnight at 4°C, and spin at 1500g for 30 minutes.In addition, PEG-it acts as a cryopreservative for concentrated virus. Lentivirus concentrated with PEG-it lasts longer in the freezer and survives multiple freeze-thaw cycles with minimal loss of titer.
Catalog No. NC9846201 Supplier System Biosciences LLC Supplier No. LV825A1
Easily concentrate lentivirus particles for ultra-high titers. Used in over three-hundred citations, PEG-it™ Virus Concentration Reagent enables easy concentration of pseudoviral particles for achieving ultra-high titers. It’s great for concentrating pseudoviral particles even from large volumes of medium by removing the need for ultracentrifugation. Simply add PEG-it to the collected medium, incubate overnight at 4°C, and spin at 1500g for 30 minutes.In addition, PEG-it acts as a cryopreservative for concentrated virus. Lentivirus concentrated with PEG-it lasts longer in the freezer and survives multiple freeze-thaw cycles with minimal loss of titer.