Learn More
Bioworld PEG Maleimide, 100 mg
Catalog No. 50199307
Encompass_Preferred
Methoxypolyethylene glycol maleimide (MeO-PEG-Mal) is used as a protein pegylation reagent. PEGylation is the process of covalently attaching polyethylene glycol (PEG) polymer chains to other molecule such as proteins.Biotin PEG MaleimideSulfhydryl reactiveWater solubleSpacer arm
Catalog No. 50-199-307 Supplier Bioworld Supplier No. 218300131
Methoxypolyethylene glycol maleimide (MeO-PEG-Mal) is used as a protein pegylation reagent. PEGylation is the process of covalently attaching polyethylene glycol (PEG) polymer chains to other molecule such as proteins.Biotin PEG MaleimideSulfhydryl reactiveWater solubleSpacer arm