Learn More
BEZWADA BIOMEDICAL LLC PEG Methacrylate, MW 4000, 5.0 GM
SDP
Catalog No. 501904503
Encompass_Preferred
Acrylate/methacrylate end functionalization enables crosslinking. Polymers used in drug delivery and various biomedical applications
Catalog No. 50-190-4503 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 74K001
Acrylate/methacrylate end functionalization enables crosslinking. Polymers used in drug delivery and various biomedical applications