Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hampton Research PEG/pH HT, 1 mL, Deep Well block format
SDP

Catalog No. NC1775194
Encompass

PEG/pH HT samples pH 3.5 - 9.6 in 0.2 M buffer.

Catalog No. NC1775194 Supplier Hampton Research Supplier No. HR2461
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG/pH HT samples pH 3.5 - 9.6 in 0.2 M buffer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.