Learn More
Medchemexpress LLC Peg2000-c-dmg | 1443687-74-8 | 98.0% | C126H249NO52 | 25 MG
SDP
Catalog No. 5000356616
Encompass_Preferred
PEG2000-C-DMG is a pegylated lipid used in the preparation of Onpattro, an investigational RNAi therapeutic agent. It is designed to reduce the production of both mutant and wild-type transthyretin by targeting the 3′ untranslated region of transthyretin mRNA.
  • Pegylated lipid
  • Used in the preparation of Onpattro
  • Targets 3′ untranslated region of transthyretin mRNA
  • Reduces production of mutant and wild-type transthyretin
Catalog No. 50-003-56616 Supplier Medchemexpress LLC Supplier No. 25MGHY145411
PEG2000-C-DMG is a pegylated lipid used in the preparation of Onpattro, an investigational RNAi therapeutic agent. It is designed to reduce the production of both mutant and wild-type transthyretin by targeting the 3′ untranslated region of transthyretin mRNA.
  • Pegylated lipid
  • Used in the preparation of Onpattro
  • Targets 3′ untranslated region of transthyretin mRNA
  • Reduces production of mutant and wild-type transthyretin