Learn More
Medchemexpress LLC PEG2000-C-DMG | 1443687-74-8 | 99.97% | C126H249NO52 | 1 ML
SDP
Catalog No. 5000355239
Encompass_Preferred
PEG2000-C-DMG is a pegylated lipid used in the preparation of an investigational RNAi therapeutic agent. This agent works by targeting the 3′ untranslated region of transthyretin mRNA, reducing the production of mutant and wild-type transthyretin.
  • Used in the preparation of an investigational RNAi therapeutic agent
  • Targets the 3′ untranslated region of transthyretin mRNA
  • Reduces production of mutant and wild-type transthyretin
  • Intended for research use only
Catalog No. 50-003-55239 Supplier Medchemexpress LLC Supplier No. HY14541110MM/1ML
PEG2000-C-DMG is a pegylated lipid used in the preparation of an investigational RNAi therapeutic agent. This agent works by targeting the 3′ untranslated region of transthyretin mRNA, reducing the production of mutant and wild-type transthyretin.
  • Used in the preparation of an investigational RNAi therapeutic agent
  • Targets the 3′ untranslated region of transthyretin mRNA
  • Reduces production of mutant and wild-type transthyretin
  • Intended for research use only