Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peg3-C4-Obn | 2381196-79-6 | 99.3% | 25 MG
SDP

Catalog No. 5000342454
Encompass_Preferred

PEG3-C4-OBn is a polyethylene glycol (PEG)-based PROTAC linker. It is utilized in the synthesis of a potent SKG3 degrader based on PROTAC.

  • Liquid appearance
  • Colorless to light yellow
  • Purity of 99.3%
  • Molecular weight of 296.40
  • Molecular formula C17H28O4
  • Used as a PROTAC linker
  • Soluble in DMSO: ≥ 100 mg/mL

Catalog No. 50-003-42454 Supplier Medchemexpress LLC Supplier No. 25MGHY130620
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG3-C4-OBn is a polyethylene glycol (PEG)-based PROTAC linker. It is utilized in the synthesis of a potent SKG3 degrader based on PROTAC.

  • Liquid appearance
  • Colorless to light yellow
  • Purity of 99.3%
  • Molecular weight of 296.40
  • Molecular formula C17H28O4
  • Used as a PROTAC linker
  • Soluble in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.