Learn More
Medchemexpress LLC Peg3-methylamine | 282551-10-4 | ≥98.0% | 163.21 | 100 MG
SDP
Catalog No. 5000348007
Encompass_Preferred
PEG3-methylamine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific protein to selectively degrade target proteins. This product is for research use only and is not sold to patients.
  • Can be used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-48007 Supplier Medchemexpress LLC Supplier No. HY140161100MG
PEG3-methylamine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific protein to selectively degrade target proteins. This product is for research use only and is not sold to patients.
  • Can be used in the synthesis of PROTACs
  • For research use only