Learn More
Medchemexpress LLC PEG3-O-CH2COOH (PROTAC Linker 8) | 51951-05-4 | ≥95.0% | 100 MG
SDP
Catalog No. 5000342920
Encompass_Preferred
PEG3-O-CH2COOH (PROTAC Linker 8) is a PEG-based PROTAC linker designed for use in the synthesis of SNIPERs. These SNIPERs recruit inhibitor of apoptosis protein (IAP) ubiquitin ligases to specifically degrade targeted proteins, offering a valuable tool for research applications.
  • PEG-based PROTAC linker
  • Used in the synthesis of SNIPERs
  • Recruits inhibitor of apoptosis protein (IAP) ubiquitin ligases
  • Designed to specifically degrade targeted proteins
  • For research use only
Catalog No. 50-003-42920 Supplier Medchemexpress LLC Supplier No. HY128804100MG
PEG3-O-CH2COOH (PROTAC Linker 8) is a PEG-based PROTAC linker designed for use in the synthesis of SNIPERs. These SNIPERs recruit inhibitor of apoptosis protein (IAP) ubiquitin ligases to specifically degrade targeted proteins, offering a valuable tool for research applications.
  • PEG-based PROTAC linker
  • Used in the synthesis of SNIPERs
  • Recruits inhibitor of apoptosis protein (IAP) ubiquitin ligases
  • Designed to specifically degrade targeted proteins
  • For research use only