Learn More
Medchemexpress LLC Peg3-o-ch2cooh (protac linker 8) | 51951-05-4 | ≥95.0% | 50 MG
SDP
Catalog No. 5000340003
Encompass_Preferred
PEG3-O-CH2COOH (PROTAC Linker 8) is a PEG-based PROTAC linker primarily used in the synthesis of SNIPERs. It can also be utilized as PROTAC linker (Compound 15b) in the synthesis of SNIPER(ER)s. These SNIPERs are designed to recruit inhibitor of apoptosis protein (IAP) ubiquitin ligases for the specific degradation of targeted proteins. This product is intended for research use only and is not for patient treatment.
  • PEG-based PROTAC linker
  • Used in the synthesis of SNIPERs
  • Utilized as PROTAC linker (Compound 15b) in the synthesis of SNIPER(ER)s
  • Recruits inhibitor of apoptosis protein (IAP) ubiquitin ligases
  • Facilitates specific degradation of targeted proteins
  • For research use only
Catalog No. 50-003-40003 Supplier Medchemexpress LLC Supplier No. HY12880450MG
Add to Cart
Add to Cart
PEG3-O-CH2COOH (PROTAC Linker 8) is a PEG-based PROTAC linker primarily used in the synthesis of SNIPERs. It can also be utilized as PROTAC linker (Compound 15b) in the synthesis of SNIPER(ER)s. These SNIPERs are designed to recruit inhibitor of apoptosis protein (IAP) ubiquitin ligases for the specific degradation of targeted proteins. This product is intended for research use only and is not for patient treatment.
  • PEG-based PROTAC linker
  • Used in the synthesis of SNIPERs
  • Utilized as PROTAC linker (Compound 15b) in the synthesis of SNIPER(ER)s
  • Recruits inhibitor of apoptosis protein (IAP) ubiquitin ligases
  • Facilitates specific degradation of targeted proteins
  • For research use only