Learn More
Medchemexpress LLC HY-Y0873 100mL Medchemexpress, PEG300 CAS:25322-68-3 Purity:>98%
SDP
Catalog No. 502029438
Encompass_Preferred
Medchemexpress, HY-Y0873 100mL PEG300 CAS:25322-68-3 PEG300 (Polyethylene glycol 300), a neutral polymer of molecular weight 300, is a water-soluble, low immunogenic and biocompatible polymer formed by repeating units of ethylene glycol[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9438 Supplier Medchemexpress LLC Supplier No. HYY0873100ML
Medchemexpress, HY-Y0873 100mL PEG300 CAS:25322-68-3 PEG300 (Polyethylene glycol 300), a neutral polymer of molecular weight 300, is a water-soluble, low immunogenic and biocompatible polymer formed by repeating units of ethylene glycol[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.