Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peg4-aminooxy-MMAF | 1415246-35-3 | 923.19 | C47H82N6O12 | 50 MG
SDP

Catalog No. 5000202269
Encompass_Preferred

PEG4-aminooxy-MMAF is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It links the antitubulin payload MMAF to a noncleavable PEG4 spacer terminated with an aminooxy functional group, enabling oxime-based conjugation to aldehyde- or ketone-containing antibodies or linkers.

  • Drug-linker conjugate for ADC research.
  • Contains an aminooxy-terminated PEG4 spacer for oxime conjugation.
  • MMAF payload provides potent antitubulin activity.
  • Molecular weight 923.19.
  • Chemical formula C47H82N6O12.
  • Available in small research pack sizes, including 50 mg.

Catalog No. 50-002-02269 Supplier Medchemexpress LLC Supplier No. HY12896850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG4-aminooxy-MMAF is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It links the antitubulin payload MMAF to a noncleavable PEG4 spacer terminated with an aminooxy functional group, enabling oxime-based conjugation to aldehyde- or ketone-containing antibodies or linkers.

  • Drug-linker conjugate for ADC research.
  • Contains an aminooxy-terminated PEG4 spacer for oxime conjugation.
  • MMAF payload provides potent antitubulin activity.
  • Molecular weight 923.19.
  • Chemical formula C47H82N6O12.
  • Available in small research pack sizes, including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.