Learn More
Medchemexpress LLC Peg4-aminooxy-MMAF | 1415246-35-3 | 99.8% | 923.19 | C47H82N6O12 | 5 MG
SDP
Catalog No. 5000202500
Encompass_Preferred
PEG4-aminooxy-MMAF is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It links the antitubulin agent MMAF to an aminooxy-functionalized PEG4 spacer to produce a noncleavable linker-payload suitable for oxime conjugation. The compound is supplied at high purity in small-milligram pack sizes for synthesis and analytical applications.
  • Used in antibody-drug conjugate research.
  • Noncleavable PEG4 spacer for stable linkage.
  • High purity suitable for analytical and synthetic applications.
  • Available in small-milligram pack sizes for laboratory use.
  • Molecular formula C47H82N6O12; molecular weight 923.19.
Catalog No. 50-002-02500 Supplier Medchemexpress LLC Supplier No. HY1289685MG
PEG4-aminooxy-MMAF is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It links the antitubulin agent MMAF to an aminooxy-functionalized PEG4 spacer to produce a noncleavable linker-payload suitable for oxime conjugation. The compound is supplied at high purity in small-milligram pack sizes for synthesis and analytical applications.
  • Used in antibody-drug conjugate research.
  • Noncleavable PEG4 spacer for stable linkage.
  • High purity suitable for analytical and synthetic applications.
  • Available in small-milligram pack sizes for laboratory use.
  • Molecular formula C47H82N6O12; molecular weight 923.19.