Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Peg4-aminooxy-MMAF | 1415246-35-3 | MFCD32201151 | 99.8% | 923.19 | C47H82N6O12 | 10 MG
SDP

Catalog No. 5000203169
Encompass_Preferred

PEG4-aminooxy-MMAF is an antibody-drug conjugate linker-drug conjugate that links the antitubulin payload monomethyl auristatin F (MMAF) to antibodies via a noncleavable PEG4 aminooxy linker for research use in ADC development. It is supplied as a white to off-white solid with high purity and is unstable in solution, so solutions should be freshly prepared.

  • High purity, suitable for research use.
  • Molecular weight 923.19.
  • Molecular formula C47H82N6O12.
  • Appearance: white to off-white solid.
  • Storage: keep at -20°C and protect from light.
  • Available in milligram-scale quantities for laboratory conjugation.

Catalog No. 50-002-03169 Supplier Medchemexpress LLC Supplier No. HY12896810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG4-aminooxy-MMAF is an antibody-drug conjugate linker-drug conjugate that links the antitubulin payload monomethyl auristatin F (MMAF) to antibodies via a noncleavable PEG4 aminooxy linker for research use in ADC development. It is supplied as a white to off-white solid with high purity and is unstable in solution, so solutions should be freshly prepared.

  • High purity, suitable for research use.
  • Molecular weight 923.19.
  • Molecular formula C47H82N6O12.
  • Appearance: white to off-white solid.
  • Storage: keep at -20°C and protect from light.
  • Available in milligram-scale quantities for laboratory conjugation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.