Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PEG4-aminooxy-MMAF | 1415246-35-3 | MFCD32201151 | 99.8% | 923.19 | C47H82N6O12 | 100 MG
SDP

Catalog No. 5000203959
Encompass_Preferred

PEG4-aminooxy-MMAF is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines the antitubulin payload MMAF with a non-cleavable PEG4 aminooxy linker. It is supplied as a solid for formulation and conjugation studies, with recommended storage at -20°C and documented solubility and in vivo formulation methods.

  • Drug-linker conjugate for ADC research.
  • Contains MMAF payload linked via a non-cleavable PEG4 aminooxy linker.
  • Molecular weight 923.19; formula C47H82N6O12.
  • CAS number 1415246-35-3.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations documented.
  • Store at -20°C and protect from light; prepare fresh solutions.

Catalog No. 50-002-03959 Supplier Medchemexpress LLC Supplier No. HY128968100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG4-aminooxy-MMAF is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines the antitubulin payload MMAF with a non-cleavable PEG4 aminooxy linker. It is supplied as a solid for formulation and conjugation studies, with recommended storage at -20°C and documented solubility and in vivo formulation methods.

  • Drug-linker conjugate for ADC research.
  • Contains MMAF payload linked via a non-cleavable PEG4 aminooxy linker.
  • Molecular weight 923.19; formula C47H82N6O12.
  • CAS number 1415246-35-3.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations documented.
  • Store at -20°C and protect from light; prepare fresh solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.