Learn More
Medchemexpress LLC Peg4-SPDP | 1305053-43-3 | 98.0% | 488.57 | C20H28N2O8S2 | 100 MG
SDP
Catalog No. 5000203458
Encompass_Preferred
PEG4-SPDP is a PEGylated, disulfide-cleavable SPDP linker used to construct antibody-drug conjugates (ADCs). It provides a PEG4 spacer to improve solubility and linker flexibility, and is designed to release payloads under reductive intracellular conditions.
  • Cleavable disulfide linker for intracellular payload release.
  • PEG4 spacer improves solubility and linker flexibility.
  • High purity supplied (~98.0%).
  • Suitable for antibody-drug conjugation and bioconjugation workflows.
Catalog No. 50-002-03458 Supplier Medchemexpress LLC Supplier No. HY126496100MG
PEG4-SPDP is a PEGylated, disulfide-cleavable SPDP linker used to construct antibody-drug conjugates (ADCs). It provides a PEG4 spacer to improve solubility and linker flexibility, and is designed to release payloads under reductive intracellular conditions.
  • Cleavable disulfide linker for intracellular payload release.
  • PEG4 spacer improves solubility and linker flexibility.
  • High purity supplied (~98.0%).
  • Suitable for antibody-drug conjugation and bioconjugation workflows.