Learn More
Medchemexpress LLC HY-Y0873A 50mL Medchemexpress, PEG400 CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0873A50ML
Encompass_Preferred
Medchemexpress, HY-Y0873A 50mL PEG400 CAS:0 PEG400 is a strongly hydrophilic polyethylene glycol used as an excellent solvent for a large number of substances. PEG400 is widely used in a variety of pharmaceutical formulations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9439