Learn More
Medchemexpress LLC PEG5-(CH2CO2H)2 | 77855-75-5 | MFCD29079379 | 99.3% | 310.30 | C12H22O9 | 1G
SDP
Catalog No. 5000235913
Encompass_Preferred
HOOCCH2O-PEG4-CH2COOH is a symmetric PEG-based dicarboxylic acid linker used as a flexible, hydrophilic spacer for the construction of bifunctional degraders and other conjugates. It provides two carboxylic acid termini for standard coupling chemistries, and its PEG4 backbone imparts solubility and conformational flexibility suited for PROTAC and homo-PROTAC synthesis.
  • Symmetric PEG4 dicarboxylic acid linker.
  • Provides carboxylic acid termini for straightforward coupling reactions.
  • PEG backbone increases solubility and flexibility.
  • Designed for use in PROTAC and homo-PROTAC synthesis.
  • High purity suitable for research applications.
  • Typically supplied in small pack sizes for lab-scale synthesis.
Catalog No. 50-002-35913 Supplier Medchemexpress LLC Supplier No. HY1247801G
HOOCCH2O-PEG4-CH2COOH is a symmetric PEG-based dicarboxylic acid linker used as a flexible, hydrophilic spacer for the construction of bifunctional degraders and other conjugates. It provides two carboxylic acid termini for standard coupling chemistries, and its PEG4 backbone imparts solubility and conformational flexibility suited for PROTAC and homo-PROTAC synthesis.
  • Symmetric PEG4 dicarboxylic acid linker.
  • Provides carboxylic acid termini for straightforward coupling reactions.
  • PEG backbone increases solubility and flexibility.
  • Designed for use in PROTAC and homo-PROTAC synthesis.
  • High purity suitable for research applications.
  • Typically supplied in small pack sizes for lab-scale synthesis.