Learn More
Medchemexpress LLC PEG5-Tos | 155130-15-7 | 96.9% | 392.46 | 1 G
SDP
Catalog No. 5000400863
Encompass_Preferred
PEG5-Tos is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-004-00863 Supplier Medchemexpress LLC Supplier No. HY234171G
PEG5-Tos is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only