Learn More
Medchemexpress LLC Peg5-tos | 155130-15-7 | 96.9% | 392.46 | 500 MG
SDP
Catalog No. 5000402102
Encompass_Preferred
PEG5-Tos is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs work by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Facilitates selective degradation of target proteins.
Catalog No. 50-004-02102 Supplier Medchemexpress LLC Supplier No. HY23417500MG
PEG5-Tos is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs work by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Facilitates selective degradation of target proteins.