Learn More
eMolecules​ PEG6-(CH2CO2H)2 | Broadpharm | 77855-76-6 | MFCD30458028 | 354.352 | C14H26O10 | 98.000 | OC(=O)COCCOCCOCCOCCOCCOCC(O)=O | 250mg | 336613443
Supplier:  eMolecules​ 336613443
Encompass_Preferred
PEG6-(CH2CO2H)2 | Broadpharm | 77855-76-6 | MFCD30458028 | 354.352 | C14H26O10 | 98.000 | OC(=O)COCCOCCOCCOCCOCCOCC(O)=O | 250mg | 336613443
Catalog No. NC1482379