Learn More
Medchemexpress LLC HY-122702 50mg Medchemexpress, PEG6-(CH2CO2H)2 CAS:77855-76-6 Purity:>98%
SDP
Catalog No. 501969637
Encompass_Preferred
Medchemexpress, HY-122702 50mg PEG6-(CH2CO2H)2 CAS:77855-76-6 PEG6-(CH2CO2H)2 is a symmetric PEG PROTAC linker, for the synthesis of Homo-PROTACs which is bivalent small-molecule dimerizers of the VHL E3 ubiquitin ligase to induce self-degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9637 Supplier Medchemexpress LLC Supplier No. HY12270250MG
Medchemexpress, HY-122702 50mg PEG6-(CH2CO2H)2 CAS:77855-76-6 PEG6-(CH2CO2H)2 is a symmetric PEG PROTAC linker, for the synthesis of Homo-PROTACs which is bivalent small-molecule dimerizers of the VHL E3 ubiquitin ligase to induce self-degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.