Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PEG8-Tos | 886469-23-4 | 250 MG
SDP

Catalog No. 5000341757
Encompass_Preferred

PEG8-Tos is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This chemical reagent is for research use only and not intended for human use.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 524.62
  • Chemical formula: C23H40O11S
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-003-41757 Supplier Medchemexpress LLC Supplier No. 250MGHY130555
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PEG8-Tos is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This chemical reagent is for research use only and not intended for human use.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 524.62
  • Chemical formula: C23H40O11S
  • Appearance: Liquid
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.