Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pega Resin

Catalog No. 761531G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-153-1G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-153-1G Supplier Sigma Aldrich Supplier No. 761531G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PEGA resins are a hydrophilic acrylamide-PEG co-polymer providing a really extensive and more uniform swelling in a wide range of solvents (from water, methanol, ethanol to tetrahydrofuran, acetonitrile and toluene). The extreme swelling volume of this support in the standard peptide synthesis solvents DCM and DMF together with their good stability make them ideally suited for the batch as well as for continuous-flow syntheses.Polyacrylamide resin possessing long polyethylene glycol spacers giving extremely high swell resin may be used for: immobilizing enzymes in aqueous solutionaffinity purification of small-molecule binding proteinssynthesis of peptide libraries with fluorescently labeled proteins

Specifications

Molecular Formula C18H38N4O8
Linear Formula C18H38N4O8
Quantity 500 mg
MDL Number MFCD03453241
Molecular Weight (g/mol) 438.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.