Learn More
TARGETMOL CHEMICALS INC PEGAMOTECAN 5MG
SDP
Catalog No. 502580794
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Pegamotecan (PEG-camptothecin) is a topoisomerase I (TOP1) inhibitor with anticancer activity and it is used in the treatment of esophageal non-small cell lung and pancreatic cancers. purity: 97%
Catalog No. 50-258-0794 Supplier TARGETMOL CHEMICALS INC Supplier No. T246105MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Pegamotecan (PEG-camptothecin) is a topoisomerase I (TOP1) inhibitor with anticancer activity and it is used in the treatment of esophageal non-small cell lung and pancreatic cancers. purity: 97%