Learn More
Medchemexpress LLC Pegaptanib sodium | 222716-86-1 | 90.5% | 50,000 Da (approximately) | 5 MG
SDP
Catalog No. 5000263327
Encompass_Preferred
Pegaptanib sodium is a PEGylated 28-mer RNA aptamer that selectively binds vascular endothelial growth factor (VEGF)-165. Supplied as the sodium salt, it is intended for research use in studies of VEGF-mediated angiogenesis and neovascular age-related macular degeneration (AMD). It provides a research-ready reagent for in vitro and in vivo experimental models.
  • Selective VEGF-165 binding suitable for anti-angiogenic research.
  • PEGylated aptamer structure provides increased molecular weight and stability.
  • Supplied as sodium salt with approximate molecular weight 50,000 Da.
  • Purity approximately 90.5% as provided by the manufacturer.
  • Available as a small research quantity (5 mg) for laboratory studies.
Catalog No. 50-002-63327 Supplier Medchemexpress LLC Supplier No. HY1095615MG
Pegaptanib sodium is a PEGylated 28-mer RNA aptamer that selectively binds vascular endothelial growth factor (VEGF)-165. Supplied as the sodium salt, it is intended for research use in studies of VEGF-mediated angiogenesis and neovascular age-related macular degeneration (AMD). It provides a research-ready reagent for in vitro and in vivo experimental models.
  • Selective VEGF-165 binding suitable for anti-angiogenic research.
  • PEGylated aptamer structure provides increased molecular weight and stability.
  • Supplied as sodium salt with approximate molecular weight 50,000 Da.
  • Purity approximately 90.5% as provided by the manufacturer.
  • Available as a small research quantity (5 mg) for laboratory studies.