Learn More
Medchemexpress LLC Pegaptanib sodium | 222716-86-1 | 90.51% | 50000 (Approximately) | 500 MG
SDP
Catalog No. 5000343619
Encompass_Preferred
Pegaptanib sodium is an RNA aptamer with polyethylene glycol modifications, directed against vascular endothelial growth factor (VEGF)-165. It can be used for the study of neovascular age-related macular degeneration (AMD).
  • Solid appearance.
  • White to off-white color.
  • Inhibits VEGF165-mediated phosphorylation of VEGFR2 and phospholipase Cγ.
  • Inhibits VEGF165-induced calcium mobilization in human umbilical vein endothelial cells.
  • Targets vascular endothelial growth factor-165.
  • Suitable for the study of neovascular age-related macular degeneration.
Catalog No. 50-003-43619 Supplier Medchemexpress LLC Supplier No. HY10956110MG
Pegaptanib sodium is an RNA aptamer with polyethylene glycol modifications, directed against vascular endothelial growth factor (VEGF)-165. It can be used for the study of neovascular age-related macular degeneration (AMD).
  • Solid appearance.
  • White to off-white color.
  • Inhibits VEGF165-mediated phosphorylation of VEGFR2 and phospholipase Cγ.
  • Inhibits VEGF165-induced calcium mobilization in human umbilical vein endothelial cells.
  • Targets vascular endothelial growth factor-165.
  • Suitable for the study of neovascular age-related macular degeneration.