Learn More
Medchemexpress LLC Pegcetacoplan (acetate) | 99.9% | (C2H4O)nC170H248N50O47S4.xC2H4O2 | 1 MG
SDP
Catalog No. 5000398603
Encompass_Preferred
Pegcetacoplan acetate is a pegylated complement C3/C3b inhibitor. It specifically binds to and inhibits C3 and C3b, thereby suppressing the complement cascade at its proximal stage. It is not known to be an inhibitor or inducer of CYP450 isoenzymes. This product can be utilized for research into complement-mediated diseases, such as paroxysmal nocturnal hemoglobinuria (PNH) and age-related macular degeneration (AMD).
  • Specifically binds to and inhibits C3 and C3b
  • Suppresses the complement cascade at its proximal stage
  • Not an inhibitor or inducer of CYP450 isoenzymes
  • Useful for research into complement-mediated diseases
  • Relevant for paroxysmal nocturnal hemoglobinuria (PNH) research
  • Relevant for age-related macular degeneration (AMD) research
Catalog No. 50-003-98603 Supplier Medchemexpress LLC Supplier No. HYP3252A1MG
Pegcetacoplan acetate is a pegylated complement C3/C3b inhibitor. It specifically binds to and inhibits C3 and C3b, thereby suppressing the complement cascade at its proximal stage. It is not known to be an inhibitor or inducer of CYP450 isoenzymes. This product can be utilized for research into complement-mediated diseases, such as paroxysmal nocturnal hemoglobinuria (PNH) and age-related macular degeneration (AMD).
  • Specifically binds to and inhibits C3 and C3b
  • Suppresses the complement cascade at its proximal stage
  • Not an inhibitor or inducer of CYP450 isoenzymes
  • Useful for research into complement-mediated diseases
  • Relevant for paroxysmal nocturnal hemoglobinuria (PNH) research
  • Relevant for age-related macular degeneration (AMD) research