Learn More
Advanced Biomatrix PEGDA-Ink bioink for BIONOVA X DLP printer 10 mL
Catalog No. 502255968
Encompass_Preferred
PEGDA-INK is 10mL of preformulated 50% PEGDA ink (700 MW) with 0.5% LAP that gels rapidly using the BIONOVA X DLP 3D Bioprinter.
This item is not returnable. View return policy
Catalog No. 50-225-5968 Supplier Advanced Biomatrix Supplier No. D16110025307
PEGDA-INK is 10mL of preformulated 50% PEGDA ink (700 MW) with 0.5% LAP that gels rapidly using the BIONOVA X DLP 3D Bioprinter.