Learn More
INTACT GENOMICS INC PE pegRNA Synthesis Kit 10 Reactions (20 µl RXN Volume)
Catalog No. 5000083016
Encompass
The IG PE pegRNA Synthesis Kit is a perfect choice for a variety of pegRNA synthesis needs. Below is a sampling of some of the key benefits. IG PE pegRNA Synthesis Kit includes everything but your target oligos (all buffers, enzymes, scaffolds). User simply needs to provide the oligos for your target: a pair of sgRNA oligo and pegRTpbs oligo generated using the template design in this manual. Rapid Workflow (Less than 1 hour) Scalable Yield (100 µl reaction)
Catalog No. 50-000-83016 Supplier INTACT GENOMICS INC Supplier No. 3404
The IG PE pegRNA Synthesis Kit is a perfect choice for a variety of pegRNA synthesis needs. Below is a sampling of some of the key benefits. IG PE pegRNA Synthesis Kit includes everything but your target oligos (all buffers, enzymes, scaffolds). User simply needs to provide the oligos for your target: a pair of sgRNA oligo and pegRTpbs oligo generated using the template design in this manual. Rapid Workflow (Less than 1 hour) Scalable Yield (100 µl reaction)