Learn More
Hampton Research PEGRx 1, 10ml
SDP
Catalog No. 500932
Encompass
Crystallization reagent kit designed to evaluate an array of polymers of varying molecular weight in a low ionic strength environment versus a wide range of pH. Polymer reagents include Polyethylene glycols, Polyethylene glycol monomethyl ethers, and Jeffamines. The molecular weight range between 200 and 20,000 is evaluated in a low ionic strength formulation. Ten different buffers are used to span the range of pH between 3.5 and 9. The primary screen variables are polymer type, polymer molecular weight, pH and low ionic strength. 10ml. Please note items from this supplier are non-returnable.
Catalog No. 50-093-2 Supplier Hampton Research Supplier No. HR2082
Crystallization reagent kit designed to evaluate an array of polymers of varying molecular weight in a low ionic strength environment versus a wide range of pH. Polymer reagents include Polyethylene glycols, Polyethylene glycol monomethyl ethers, and Jeffamines. The molecular weight range between 200 and 20,000 is evaluated in a low ionic strength formulation. Ten different buffers are used to span the range of pH between 3.5 and 9. The primary screen variables are polymer type, polymer molecular weight, pH and low ionic strength. 10ml. Please note items from this supplier are non-returnable.