Learn More
Hampton Research HR2-982-01/PEGRx 1 #01 (A1), 185 ml - 0.1 M Citric acid pH 3.5, 34% v/v Polyethylene glycol 200
SDP
Catalog No. 502294
Encompass
HR2-982-01/PEGRx 1 #01 (A1), 185 ml - 0.1 M Citric acid pH 3.5, 34% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-229-4 Supplier Hampton Research Supplier No. HR298201
HR2-982-01/PEGRx 1 #01 (A1), 185 ml - 0.1 M Citric acid pH 3.5, 34% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.