Learn More
Hampton Research HR2-982-04/PEGRx 1 #04 (A4), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 30% v/v Polyethylene glycol 300
SDP
Catalog No. 502297
Encompass
HR2-982-04/PEGRx 1 #04 (A4), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 30% v/v Polyethylene glycol 300. Please note items from this supplier are non-returnable.
Catalog No. 50-229-7 Supplier Hampton Research Supplier No. HR298204
HR2-982-04/PEGRx 1 #04 (A4), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 30% v/v Polyethylene glycol 300. Please note items from this supplier are non-returnable.