Learn More
Hampton Research HR2-982-06/PEGRx 1 #06 (A6), 185 ml - 0.1 M BICINE pH 8.5, 20% v/v Polyethylene glycol 300
SDP
Catalog No. 502299
Encompass
HR2-982-06/PEGRx 1 #06 (A6), 185 ml - 0.1 M BICINE pH 8.5, 20% v/v Polyethylene glycol 300. Please note items from this supplier are non-returnable.
Catalog No. 50-229-9 Supplier Hampton Research Supplier No. HR298206
HR2-982-06/PEGRx 1 #06 (A6), 185 ml - 0.1 M BICINE pH 8.5, 20% v/v Polyethylene glycol 300. Please note items from this supplier are non-returnable.