Learn More
Hampton Research HR2-982-07/PEGRx 1 #07 (A7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.0, 15% v/v Polyethylene glycol 400
SDP
Catalog No. 502300
Encompass
HR2-982-07/PEGRx 1 #07 (A7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.0, 15% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-230-0 Supplier Hampton Research Supplier No. HR298207
HR2-982-07/PEGRx 1 #07 (A7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.0, 15% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.