Learn More
Hampton Research HR2-982-08/PEGRx 1 #08 (A8), 185 ml - 0.1 M MES monohydrate pH 6.0, 22% v/v Polyethylene glycol 400
SDP
Catalog No. 502301
Encompass
HR2-982-08/PEGRx 1 #08 (A8), 185 ml - 0.1 M MES monohydrate pH 6.0, 22% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-230-1 Supplier Hampton Research Supplier No. HR298208
HR2-982-08/PEGRx 1 #08 (A8), 185 ml - 0.1 M MES monohydrate pH 6.0, 22% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.