Learn More
Hampton Research HR2-982-09/PEGRx 1 #09 (A9), 185 ml - 0.1 M Tris pH 8.0, 30% v/v Polyethylene glycol 400
SDP
Catalog No. 502302
Encompass
HR2-982-09/PEGRx 1 #09 (A9), 185 ml - 0.1 M Tris pH 8.0, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-230-2 Supplier Hampton Research Supplier No. HR298209
HR2-982-09/PEGRx 1 #09 (A9), 185 ml - 0.1 M Tris pH 8.0, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.