Learn More
Hampton Research HR2-982-15/PEGRx 1 #15 (B3), 185 ml - 0.1 M Tris pH 8.0, 30% v/v Jeffamine M-600 pH 7.0
SDP
Catalog No. 502308
Encompass
HR2-982-15/PEGRx 1 #15 (B3), 185 ml - 0.1 M Tris pH 8.0, 30% v/v Jeffamine M-600 pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-230-8 Supplier Hampton Research Supplier No. HR298215
HR2-982-15/PEGRx 1 #15 (B3), 185 ml - 0.1 M Tris pH 8.0, 30% v/v Jeffamine M-600 pH 7.0. Please note items from this supplier are non-returnable.