Learn More
Hampton Research HR2-982-16/PEGRx 1 #16 (B4), 185 ml - 0.1 M Citric acid pH 3.5, 14% w/v Polyethylene glycol 1,000
SDP
Catalog No. 502309
Encompass
HR2-982-16/PEGRx 1 #16 (B4), 185 ml - 0.1 M Citric acid pH 3.5, 14% w/v Polyethylene glycol 1,000. Please note items from this supplier are non-returnable.
Catalog No. 50-230-9 Supplier Hampton Research Supplier No. HR298216
HR2-982-16/PEGRx 1 #16 (B4), 185 ml - 0.1 M Citric acid pH 3.5, 14% w/v Polyethylene glycol 1,000. Please note items from this supplier are non-returnable.