Learn More
Hampton Research HR2-982-17/PEGRx 1 #17 (B5), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 22% w/v Polyethylene glycol 1,000
SDP
Catalog No. 502310
Encompass
HR2-982-17/PEGRx 1 #17 (B5), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 22% w/v Polyethylene glycol 1,000. Please note items from this supplier are non-returnable.
Catalog No. 50-231-0 Supplier Hampton Research Supplier No. HR298217
HR2-982-17/PEGRx 1 #17 (B5), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 22% w/v Polyethylene glycol 1,000. Please note items from this supplier are non-returnable.