Learn More
Hampton Research HR2-982-19/PEGRx 1 #19 (B7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 30% w/v Polyethylene glycol 1,500
SDP
Catalog No. 502312
Encompass
HR2-982-19/PEGRx 1 #19 (B7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 30% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.
Catalog No. 50-231-2 Supplier Hampton Research Supplier No. HR298219
HR2-982-19/PEGRx 1 #19 (B7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 30% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.