Learn More
Hampton Research HR2-982-21/PEGRx 1 #21 (B9), 185 ml - 0.1 M BICINE pH 8.5, 15% w/v Polyethylene glycol 1,500
SDP
Catalog No. 502314
Encompass
HR2-982-21/PEGRx 1 #21 (B9), 185 ml - 0.1 M BICINE pH 8.5, 15% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.
Catalog No. 50-231-4 Supplier Hampton Research Supplier No. HR298221
HR2-982-21/PEGRx 1 #21 (B9), 185 ml - 0.1 M BICINE pH 8.5, 15% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.