Learn More
Hampton Research HR2-982-23/PEGRx 1 #23 (B11), 185 ml - 0.1 M MES monohydrate pH 6.0, 20% w/v Polyethylene glycol monomethyl ether 2,000
SDP
Catalog No. 502316
Encompass
HR2-982-23/PEGRx 1 #23 (B11), 185 ml - 0.1 M MES monohydrate pH 6.0, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-231-6 Supplier Hampton Research Supplier No. HR298223
HR2-982-23/PEGRx 1 #23 (B11), 185 ml - 0.1 M MES monohydrate pH 6.0, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.