Learn More
Hampton Research HR2-982-25/PEGRx 1 #25 (C1), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 30% v/v Jeffamine ED-2001 pH 7.0
SDP
Catalog No. 502318
Encompass
HR2-982-25/PEGRx 1 #25 (C1), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 30% v/v Jeffamine ED-2001 pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-231-8 Supplier Hampton Research Supplier No. HR298225
HR2-982-25/PEGRx 1 #25 (C1), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 30% v/v Jeffamine ED-2001 pH 7.0. Please note items from this supplier are non-returnable.