Learn More
Hampton Research HR2-982-28/PEGRx 1 #28 (C4), 185 ml - 0.1 M Citric acid pH 3.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502321
Encompass
HR2-982-28/PEGRx 1 #28 (C4), 185 ml - 0.1 M Citric acid pH 3.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-232-1 Supplier Hampton Research Supplier No. HR298228
HR2-982-28/PEGRx 1 #28 (C4), 185 ml - 0.1 M Citric acid pH 3.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.