Learn More
Hampton Research HR2-982-29/PEGRx 1 #29 (C5), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 18% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502322
Encompass
HR2-982-29/PEGRx 1 #29 (C5), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 18% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-232-2 Supplier Hampton Research Supplier No. HR298229
HR2-982-29/PEGRx 1 #29 (C5), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 18% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.