Learn More
Hampton Research HR2-982-35/PEGRx 1 #35 (C11), 185 ml - 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502328
Encompass
HR2-982-35/PEGRx 1 #35 (C11), 185 ml - 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-232-8 Supplier Hampton Research Supplier No. HR298235
HR2-982-35/PEGRx 1 #35 (C11), 185 ml - 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.