Learn More
Hampton Research HR2-982-38/PEGRx 1 #38 (D2), 185 ml - 0.1 M Imidazole pH 7.0, 20% w/v Polyethylene glycol 6,000
SDP
Catalog No. 502331
Encompass
HR2-982-38/PEGRx 1 #38 (D2), 185 ml - 0.1 M Imidazole pH 7.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-233-1 Supplier Hampton Research Supplier No. HR298238
HR2-982-38/PEGRx 1 #38 (D2), 185 ml - 0.1 M Imidazole pH 7.0, 20% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.